accessibility menu, dialog, popup

UserName

HNOCl

CAS RN 13466-35-8

IUPAC Name: 3-chloro-1,2-dihydropyridin-2-one
Synonyms: 3-chloro-1H-pyridin-2-one
Molecular Weight (g/mol): 129.54
Molecular Formula: C5H4ClNO
InChi Key: XTYNIPUFKBBALX-UHFFFAOYSA-N
SMILES: ClC1=CC=CNC1=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)