missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-fluorobenzyl Bromide 96.0+%, TCI America™
Supplier: TCI America C21185G
Specifications
| 3-Chloro-2-fluorobenzyl Bromide | |
| White-Yellow | |
| C7H5BrClF | |
| 5 g | |
| 3-chloro-2-fluorobenzyl bromide, 1-bromomethyl-3-chloro-2-fluorobenzene, 2-fluoro-3-chlorobenzyl bromide, 3-chloro-2-fluorobenzylbromide, 3-chloro-2-fluoro benzyl bromide, alpha-bromo-3-chloro-2-fluorotoluene, alpha-bromo-2-fluoro-3-chlorotoluene, benzene, 1-bromomethyl-3-chloro-2-fluoro, 1-bromomethyl-3-chloro-2-fluoro-benzene | |
| FC1=C(Cl)C=CC=C1CBr | |
| 223.47 | |
| 223.47 | |
| Crystals |
| 85070-47-9 | |
| 126°C | |
| MFCD00042503 | |
| 3261 | |
| HYILLTADABKYHO-UHFFFAOYSA-N | |
| 1-(bromomethyl)-3-chloro-2-fluorobenzene | |
| 522827 | |
| ≥96.0% (GC,T) |
Chemical Identifiers
| 85070-47-9 | |
| 223.47 | |
| HYILLTADABKYHO-UHFFFAOYSA-N | |
| 522827 | |
| FC1=C(Cl)C=CC=C1CBr |
| C7H5BrClF | |
| MFCD00042503 | |
| 3-chloro-2-fluorobenzyl bromide, 1-bromomethyl-3-chloro-2-fluorobenzene, 2-fluoro-3-chlorobenzyl bromide, 3-chloro-2-fluorobenzylbromide, 3-chloro-2-fluoro benzyl bromide, alpha-bromo-3-chloro-2-fluorotoluene, alpha-bromo-2-fluoro-3-chlorotoluene, benzene, 1-bromomethyl-3-chloro-2-fluoro, 1-bromomethyl-3-chloro-2-fluoro-benzene | |
| 1-(bromomethyl)-3-chloro-2-fluorobenzene |
Safety and Handling
TSCA : No