missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. c255325g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C255325G 25 g
C25535G 5 g
2 options

Catalog No. C255325G

Supplier: TCI America C255325G

Only null left

Chemical Identifiers

CAS 351003-48-0
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD03094234
InChI Key LIRQNYQIFFLGIE-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorobenzenesulphonyl chloride, 3-chloro-2-fluorobenzene-1-sulfonyl chloride, 3-chloro-2-fluorobenzenesulphonylchloride, 3-chloro-2-fluorophenylsulfonyl chloride, 3-chloro-2-fluorobenzensulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-fluoro, pubchem8446, acmc-209vns, 3-chloro-2-fluoro-benzenesulfonyl chloride, ksc222a3d
PubChem CID 2778321
IUPAC Name 3-chloro-2-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)F)S(=O)(=O)Cl

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 229.05
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-fluorobenzenesulfonyl Chloride