accessibility menu, dialog, popup

UserName

CAS RN 351003-48-0

IUPAC Name: 3-chloro-2-fluorobenzene-1-sulfonyl chloride
Synonyms: 3-chloro-2-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 229.05
Molecular Formula: C6H3Cl2FO2S
InChi Key: LIRQNYQIFFLGIE-UHFFFAOYSA-N
SMILES: FC1=C(C=CC=C1Cl)S(Cl)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results