missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-fluorobenzaldehyde 95.0+%, TCI America™
$178.95 - $533.13
Chemical Identifiers
| CAS | 85070-48-0 |
|---|---|
| Molecular Formula | C7H4ClFO |
| Molecular Weight (g/mol) | 158.56 |
| MDL Number | MFCD01631571 |
| InChI Key | YAOZCMANASAVFN-UHFFFAOYSA-N |
| Synonym | 2-fluoro-3-chlorobenzaldehyde, 2-fluoro-3-chloro-benzaldehyde, 3-chloro-2-fluoro-benzaldehyde, benzaldehyde, 3-chloro-2-fluoro, pubchem1408, acmc-209q3u, 2-fluoro chloro benzaldehyde, ksc496e9b, 3-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003987 |
| PubChem CID | 736335 |
| IUPAC Name | 3-chloro-2-fluorobenzaldehyde |
| SMILES | FC1=C(Cl)C=CC=C1C=O |
Chemical Identifiers
| 85070-48-0 | |
| 158.56 | |
| YAOZCMANASAVFN-UHFFFAOYSA-N | |
| 736335 | |
| FC1=C(Cl)C=CC=C1C=O |
| C7H4ClFO | |
| MFCD01631571 | |
| 2-fluoro-3-chlorobenzaldehyde, 2-fluoro-3-chloro-benzaldehyde, 3-chloro-2-fluoro-benzaldehyde, benzaldehyde, 3-chloro-2-fluoro, pubchem1408, acmc-209q3u, 2-fluoro chloro benzaldehyde, ksc496e9b, 3-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003987 | |
| 3-chloro-2-fluorobenzaldehyde |
Specifications
| 85070-48-0 | |
| 214°C | |
| MFCD01631571 | |
| 2-fluoro-3-chlorobenzaldehyde, 2-fluoro-3-chloro-benzaldehyde, 3-chloro-2-fluoro-benzaldehyde, benzaldehyde, 3-chloro-2-fluoro, pubchem1408, acmc-209q3u, 2-fluoro chloro benzaldehyde, ksc496e9b, 3-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003987 | |
| FC1=C(Cl)C=CC=C1C=O | |
| 158.56 | |
| 158.56 | |
| Liquid |
| Yellow | |
| C7H4ClFO | |
| 5 g | |
| YAOZCMANASAVFN-UHFFFAOYSA-N | |
| 3-chloro-2-fluorobenzaldehyde | |
| 736335 | |
| ≥95.0% (GC) | |
| 3-Chloro-2-fluorobenzaldehyde |
Safety and Handling
TSCA : No