missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-fluoroaniline 98.0+%, TCI America™
$32.72 - $67.38
Chemical Identifiers
| CAS | 2106-04-9 |
|---|---|
| Molecular Formula | C6H5ClFN |
| Molecular Weight (g/mol) | 145.56 |
| MDL Number | MFCD00069415 |
| InChI Key | XWBTZHDDWRNOQH-UHFFFAOYSA-N |
| Synonym | 2-fluoro-3-chloroaniline, benzenamine, 3-chloro-2-fluoro, 3-chloro-2-fluorophenylamine, pubchem1529, 3,4-chlorofluoroaniline, 3-chloro-2 fluoroaniline, 2-fluoro-3-chloro-aniline, 3-chloro-2-fluoro aniline, 3-chloro-2-fluoro-aniline, ksc493k4l |
| PubChem CID | 75014 |
| IUPAC Name | 3-chloro-2-fluoroaniline |
| SMILES | C1=CC(=C(C(=C1)Cl)F)N |
Chemical Identifiers
| 2106-04-9 | |
| 145.56 | |
| XWBTZHDDWRNOQH-UHFFFAOYSA-N | |
| 75014 | |
| C1=CC(=C(C(=C1)Cl)F)N |
| C6H5ClFN | |
| MFCD00069415 | |
| 2-fluoro-3-chloroaniline, benzenamine, 3-chloro-2-fluoro, 3-chloro-2-fluorophenylamine, pubchem1529, 3,4-chlorofluoroaniline, 3-chloro-2 fluoroaniline, 2-fluoro-3-chloro-aniline, 3-chloro-2-fluoro aniline, 3-chloro-2-fluoro-aniline, ksc493k4l | |
| 3-chloro-2-fluoroaniline |
Specifications
| 2106-04-9 | |
| 5 g | |
| C6H5ClFN | |
| 2-fluoro-3-chloroaniline, benzenamine, 3-chloro-2-fluoro, 3-chloro-2-fluorophenylamine, pubchem1529, 3,4-chlorofluoroaniline, 3-chloro-2 fluoroaniline, 2-fluoro-3-chloro-aniline, 3-chloro-2-fluoro aniline, 3-chloro-2-fluoro-aniline, ksc493k4l | |
| C1=CC(=C(C(=C1)Cl)F)N | |
| 145.56 | |
| 145.56 | |
| 3-Chloro-2-fluoroaniline |
| 99°C | |
| Clear Liquid at 20°C | |
| MFCD00069415 | |
| XWBTZHDDWRNOQH-UHFFFAOYSA-N | |
| 3-chloro-2-fluoroaniline | |
| 75014 | |
| ≥98.0% (GC) |
Safety and Handling
TSCA : Yes