missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoroaniline, 97%

CAS: 2106-04-9 | C8H11Cl2N5 | 248.11 g/mol

$50.48 - $381.30

Chemical Identifiers

CAS 2106-04-9
MDL Number MFCD00069415
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5114406
View Documents
Thermo Scientific Chemicals
H5114406
5 g
Each for $50.48
Only null left
 
AAH5114422
View Documents
Thermo Scientific Chemicals
H5114422
100 g
Each for $381.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2106-04-9
MFCD00069415

Specifications

2106-04-9
214°C
Characteristic
C6H5ClFN
UN2810
XWBTZHDDWRNOQH-UHFFFAOYSA-N
3-chloro-2-fluoroaniline
75014
97%
1.324
5 g
1.564
MFCD00069415
2-fluoro-3-chloroaniline, benzenamine, 3-chloro-2-fluoro, 3-chloro-2-fluorophenylamine, pubchem1529, 3,4-chlorofluoroaniline, 3-chloro-2 fluoroaniline, 2-fluoro-3-chloro-aniline, 3-chloro-2-fluoro aniline, 3-chloro-2-fluoro-aniline, ksc493k4l
C1=CC(=C(C(=C1)Cl)F)N
145.56
145.56
3-Chloro-2-fluoroaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only