missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoro-5-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog No. p-7057067
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66400MD 250 mg
AAH6640003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66400MD Supplier Thermo Scientific Chemicals Supplier No. H66400MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 209328-72-3
Molecular Formula C5H3ClFNO
Molecular Weight (g/mol) 147.533
MDL Number MFCD08061311
InChI Key IETLNKVWDZYCPN-UHFFFAOYSA-N
PubChem CID 22451555
IUPAC Name 5-chloro-6-fluoropyridin-3-ol
SMILES C1=C(C=NC(=C1Cl)F)O

Specifications

Quantity 250 mg
Beilstein 13815075
Synonym 3-chloro-2-fluoro-5-hydroxypyridine, 3-pyridinol, 5-chloro-6-fluoro, 5-chloro-6-fluoro-3-pyridinol, pubchem6262, 5-chloro-6-fluoro-pyridin-3-ol, 3-pyridinol,5-chloro-6-fluoro, 3-pyridinol, 5-chloro-6-fluoro-9ci, 2-fluoro-3-chloro-5-hydroxypyridine
Formula Weight 147.53
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Chloro-2-fluoro-5-hydroxypyridine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only