Learn More
3-chloro-2-chloromethyl-1-propene, 96%
CAS: 1871-57-4 | C4H6Cl2 | 124.99 g/mol
Supplier: Thermo Scientific Chemicals 277070250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-chloro-2-chloromethyl-1-propene | |
| 95.0 | |
| 14°C | |
| 1.0800g/mL | |
| 36°C | |
| 95% min. (GC) | |
| C4H6Cl2 | |
| H2C=C(CH2Cl)2 | |
| 25 g | |
| 1.08 | |
| Solubility in water: insoluble | |
| ClCC(=C)CCl | |
| 124.99 | |
| 125 | |
| Liquid |
| 1871-57-4 | |
| 100.0 | |
| Colorless to Yellow | |
| 138°C to 140°C | |
| Authentic | |
| Glass bottle | |
| 1.4750 to 1.479 | |
| MFCD00000952 | |
| 01, II, 181 | |
| 3-chloro-2-chloromethyl prop-1-ene, 3-chloro-2-chloromethyl-1-propene, 1,1-bis chloromethyl ethylene, methallyl dichloride, 1-propene, 3-chloro-2-chloromethyl, 2-chloromethyl-3-chloropropene, 1,3-dichloro-2-methylenepropane, 3-chloro-2-chloromethyl propene, 2-methylene-1,3-dichloropropane, 2-methylenepropane-1,3-dichloride | |
| XJFZOSUFGSANIF-UHFFFAOYSA-N | |
| 3-chloro-2-(chloromethyl)prop-1-ene | |
| 15859 | |
| 96% |
Chemical Identifiers
| 1871-57-4 | |
| 124.99 | |
| XJFZOSUFGSANIF-UHFFFAOYSA-N | |
| 15859 | |
| ClCC(=C)CCl |
| C4H6Cl2 | |
| MFCD00000952 | |
| 3-chloro-2-chloromethyl prop-1-ene, 3-chloro-2-chloromethyl-1-propene, 1,1-bis chloromethyl ethylene, methallyl dichloride, 1-propene, 3-chloro-2-chloromethyl, 2-chloromethyl-3-chloropropene, 1,3-dichloro-2-methylenepropane, 3-chloro-2-chloromethyl propene, 2-methylene-1,3-dichloropropane, 2-methylenepropane-1,3-dichloride | |
| 3-chloro-2-(chloromethyl)prop-1-ene |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes serious eye irritation.
Fatal if inhaled.
Very toxic to aquatic life.
Flammable liquid and vapor.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 217-489-3
RUO – Research Use Only