missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-butanone 95.0+%, TCI America™

Catalog No. p-7124258
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C017025ML 25 mL
C0170500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C017025ML Supplier TCI America Supplier No. C017025ML
Only null left

Chemical Identifiers

CAS 4091-39-8
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
MDL Number MFCD00000865
InChI Key OIMRLHCSLQUXLL-UHFFFAOYSA-N
Synonym 3-chloro-2-butanone, 2-butanone, 3-chloro, 3-chlorobutanone, 1-acetyl-1-chloroethane, 1-chloroethyl methyl ketone, 2-chloro-3-butanone, 2-butanone, 3-chloro-, +, chloro-mek, 3-chloro-2-butanon, 3-chlorobutan-2-on
PubChem CID 20026
IUPAC Name 3-chlorobutan-2-one
SMILES CC(C(=O)C)Cl

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 mL
UN Number 1224
Formula Weight 106.55
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-butanone
EINECSNumber (2)-0578
RTECSNumber EL7062000
TSCA Yes