accessibility menu, dialog, popup

UserName

CAS RN 4091-39-8

IUPAC Name: 3-chlorobutan-2-one
Synonyms: 3-chloro-2-butanone
Molecular Weight (g/mol): 106.55
Molecular Formula: C4H7ClO
InChi Key: OIMRLHCSLQUXLL-UHFFFAOYNA-N
SMILES: CC(Cl)C(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)