missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-butanone, 96%

Catalog No. AAL1415836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1415836 500 g
AAL1415822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1415836 Supplier Thermo Scientific Chemicals Supplier No. L1415836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4091-39-8 | C4H7ClO | 106.549 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4091-39-8
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
MDL Number MFCD00000865
InChI Key OIMRLHCSLQUXLL-UHFFFAOYSA-N
Synonym 3-chloro-2-butanone, 2-butanone, 3-chloro, 3-chlorobutanone, 1-acetyl-1-chloroethane, 1-chloroethyl methyl ketone, 2-chloro-3-butanone, 2-butanone, 3-chloro-, +, chloro-mek, 3-chloro-2-butanon, 3-chlorobutan-2-on
PubChem CID 20026
IUPAC Name 3-chlorobutan-2-one
SMILES CC(C(=O)C)Cl

Specifications

Melting Point <-60°C
Density 1.055
Boiling Point 113°C to 116°C
Flash Point 28°C (82°F)
Refractive Index 1.421
Quantity 500 g
UN Number UN1224
Beilstein 385637
Formula Weight 106.55
Percent Purity 96%
Chemical Name or Material 3-Chloro-2-butanone
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 223-834-9
RTECSNumber EL7062000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only