missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,5,6-trifluoropyridine, 98+%

Catalog No. p-7047638
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1954306 5 g
AAL1954314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1954306 Supplier Thermo Scientific Chemicals Supplier No. L1954306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2879-42-7 | C5HClF3N | 167.52 g/mol

3-Chloro-2,5,6-trifluoropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-2,5,6-trifluoropyridine is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2879-42-7
Molecular Formula C5HClF3N
Molecular Weight (g/mol) 167.52
MDL Number MFCD03001160
InChI Key OROKHXKJSDPRET-UHFFFAOYSA-N
PubChem CID 2783252
IUPAC Name 3-chloro-2,5,6-trifluoropyridine
SMILES FC1=CC(Cl)=C(F)N=C1F

Specifications

Boiling Point 132°C to 134°C
Flash Point >100°C (212°F)
Odor Unpleasant
Refractive Index 1.456
Quantity 5 g
UN Number UN1760
Beilstein 1424722
Synonym pyridine,3-chloro-2,5,6-trifluoro, pyridine, 3-chloro-2,5,6-trifluoro, 3-chloro-2,5,6-trifluoro-pyridine, acmc-1cekn, 2,3,6-trifluoro-5-chloropyridine, 5-chloro-2,3,6-trifluoropyridine, 3-chloranyl-2,5,6-tris fluoranyl pyridine
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 167.52
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-2,5,6-trifluoropyridine
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only