accessibility menu, dialog, popup

UserName

FClFNF

CAS RN 2879-42-7

IUPAC Name: 3-chloro-2,5,6-trifluoropyridine
Synonyms: 3-chloro-2,5,6-trifluoropyridine
Molecular Weight (g/mol): 167.52
Molecular Formula: C5HClF3N
InChi Key: OROKHXKJSDPRET-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=C(F)N=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results