missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4,5-trifluorobenzoic Acid 98.0+%, TCI America™

Catalog No. C22615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22615G 5 g
C226125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22615G Supplier TCI America Supplier No. C22615G
Only null left

Chemical Identifiers

CAS 101513-77-3
Molecular Formula C7H2ClF3O2
Molecular Weight (g/mol) 210.536
MDL Number MFCD00153101
InChI Key KBESHLYCSZINAJ-UHFFFAOYSA-N
Synonym 2,4,5-trifluoro-3-chlorobenzoic acid, 3-chloro-2,4,5-trifluoro-benzoic acid, 3-chloro-2,4,5-trifluorobenzoicacid, benzoic acid, 3-chloro-2,4,5-trifluoro, pubchem1376, acmc-1bvtw, ksc502q6n, 3-chloro-2,4,5trifluorobenzoic acid, 3-chloro-2,4,5-trilfuorobenzoic acid, 3-carboxy-2,5,6-trifluorochlorobenzene
PubChem CID 2734269
IUPAC Name 3-chloro-2,4,5-trifluorobenzoic acid
SMILES C1=C(C(=C(C(=C1F)F)Cl)F)C(=O)O

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 5 g
Formula Weight 210.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2,4,5-trifluorobenzoic Acid