missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4,5,6-tetrafluoropyridine 98.0+%, TCI America™

Catalog No. p-7125053
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
C15495G 5 g
C154925G 25 g
2 options

Catalog No. C15495G

Supplier: TCI America C15495G

Only null left

Chemical Identifiers

CAS 1735-84-8
Molecular Formula C5ClF4N
Molecular Weight (g/mol) 185.51
MDL Number MFCD00010624
InChI Key UXUZMYHSICIOQT-UHFFFAOYSA-N
Synonym pyridine, 3-chloro-2,4,5,6-tetrafluoro, 3-chlorotetrafluoropyridine, 2-chloro-2,4,5,6-tetrafluoropyridine, pubchem9226, 2,3,4,6-tetrafluoro-5-chloropyridine, 5-chloro-2,3,4,6-tetrafluoropyridine, 3-chloro-2,4,5,6-tetrafluoro-pyridine, pyridine,3-chloro-2,4,5,6-tetrafluoro, 3-chloro-2,4,5,6-tetrafluoropyridine, 3-chloranyl-2,4,5,6-tetrakis fluoranyl pyridine
PubChem CID 137183
IUPAC Name 3-chloro-2,4,5,6-tetrafluoropyridine
SMILES FC1=NC(F)=C(Cl)C(F)=C1F

Specifications

Color Yellow
Boiling Point 71°C
Quantity 5 g
Formula Weight 185.51
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluoropyridine