missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-propanol 98.0+%, TCI America™

Catalog No. p-7124323
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C027025G 25 g
C0270100G 100 g
C0270500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C027025G Supplier TCI America Supplier No. C027025G

Chemical Identifiers

CAS 627-30-5
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.538
MDL Number MFCD00002943
InChI Key LAMUXTNQCICZQX-UHFFFAOYSA-N
Synonym 3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
PubChem CID 12313
IUPAC Name 3-chloropropan-1-ol
SMILES C(CO)CCl

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
UN Number 2849
Formula Weight 94.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-1-propanol
EINECSNumber (2)-2002
RTECSNumber UA8930000
TSCA Yes