missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-1-propanol, 98%, stab.
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Soluble in water at 20°C 300g/L.
Notes
Store away from oxidizing agents and bases. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
It is used as a pharmaceutical intermediate.
Solubility
Soluble in water at 20°C 300g/L.
Notes
Store away from oxidizing agents and bases. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
Specifications
| Melting Point | -20°C |
| Density | 1.131 |
| pH | 3 to 4 |
| Boiling Point | 160°C to 162°C |
| Flash Point | 73°C (163°F) |
| Odor | Pleasant |
| Refractive Index | 1.445 |
| Quantity | 25 g |
| UN Number | UN2849 |
| Beilstein | 773655 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 3-CHLOROPROPANOL-1 |
| EINECSNumber | 210-992-9 |
| RTECSNumber | UA8930000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only