missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-propanol, 98%, stab.

Catalog No. AAA1687114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1687114 25 g
AAA1687122 100 g
AAA1687136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1687114 Supplier Thermo Scientific Chemicals Supplier No. A1687114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 627-30-5 | C3H7ClO | 94.538 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 20°C 300g/L.

Notes
Store away from oxidizing agents and bases. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 627-30-5
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.538
MDL Number MFCD00002943
InChI Key LAMUXTNQCICZQX-UHFFFAOYSA-N
Synonym 3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
PubChem CID 12313
IUPAC Name 3-chloropropan-1-ol
SMILES C(CO)CCl

Specifications

Melting Point -20°C
Density 1.131
pH 3 to 4
Boiling Point 160°C to 162°C
Flash Point 73°C (163°F)
Odor Pleasant
Refractive Index 1.445
Quantity 25 g
UN Number UN2849
Beilstein 773655
Solubility Information Soluble in water at 20°C 300g/L.
Formula Weight 94.54
Percent Purity 98%
Chemical Name or Material 3-Chloro-1-propanol
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 3-CHLOROPROPANOL-1
EINECSNumber 210-992-9
RTECSNumber UA8930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only