missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-butyne 98.0+%, TCI America™

Catalog No. C11951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11951G 1 g
C11955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11951G Supplier TCI America Supplier No. C11951G
Only null left

Chemical Identifiers

CAS 21020-24-6
Molecular Formula C4H5Cl
Molecular Weight (g/mol) 88.53
MDL Number MFCD00143109
InChI Key PZFBULOUMNPBFA-UHFFFAOYNA-N
PubChem CID 30443
IUPAC Name 3-chlorobut-1-yne
SMILES CC(Cl)C#C

Specifications

Color Yellow
Boiling Point 68°C
Quantity 1 g
UN Number 1993
Formula Weight 88.53
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-1-butyne