accessibility menu, dialog, popup

UserName

CH3ClHC

CAS RN 21020-24-6

IUPAC Name: 3-chlorobut-1-yne
Synonyms: 3-chloro-1-butyne
Molecular Weight (g/mol): 88.53
Molecular Formula: C4H5Cl
InChi Key: PZFBULOUMNPBFA-UHFFFAOYNA-N
SMILES: CC(Cl)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results