missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-1,2-propanediol 98.0+%, TCI America™
Supplier: TCI America C026825G
Specifications
| 3-Chloro-1,2-propanediol | |
| -40°C | |
| C3H7ClO2 | |
| 25 g | |
| 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin | |
| OCC(O)CCl | |
| 110.54 | |
| CHEBI:18721 | |
| ≥98.0% (GC) |
| 96-24-2 | |
| Yellow | |
| MFCD00004712 | |
| 2689 | |
| SSZWWUDQMAHNAQ-UHFFFAOYNA-N | |
| 3-chloropropane-1,2-diol | |
| 7290 | |
| 110.54 | |
| Liquid |
Chemical Identifiers
| 96-24-2 | |
| 110.54 | |
| SSZWWUDQMAHNAQ-UHFFFAOYNA-N | |
| 7290 | |
| 3-chloropropane-1,2-diol |
| C3H7ClO2 | |
| MFCD00004712 | |
| 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin | |
| CHEBI:18721 | |
| OCC(O)CCl |
Safety and Handling
EINECSNumber : (2)-2441
RTECSNumber : TY4025000
TSCA : Yes