missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-propanediol 98.0+%, TCI America™

Catalog No. p-7124321
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C026825G 25 g
C0268500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C026825G Supplier TCI America Supplier No. C026825G
Only null left

Chemical Identifiers

CAS 96-24-2
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.54
MDL Number MFCD00004712
InChI Key SSZWWUDQMAHNAQ-UHFFFAOYNA-N
Synonym 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
PubChem CID 7290
ChEBI CHEBI:18721
IUPAC Name 3-chloropropane-1,2-diol
SMILES OCC(O)CCl

Specifications

Melting Point -40°C
Color Yellow
Quantity 25 g
UN Number 2689
Formula Weight 110.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-1,2-propanediol
EINECSNumber (2)-2441
RTECSNumber TY4025000
TSCA Yes