Learn More
(+/-)-3-Chloro-1,2-propanediol, 98%
CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol
Supplier: Thermo Scientific Chemicals A1614336
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (+/-)-3-Chloro-1,2-propanediol | |
| -40°C | |
| 213°C | |
| C3H7ClO2 | |
| MFCD00004712 | |
| UN2689 | |
| Hygroscopic | |
| 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin | |
| OCC(O)CCl | |
| 110.54 | |
| CHEBI:18721 | |
| 98% |
| 96-24-2 | |
| 1.322 | |
| 137°C (278°F) | |
| 1.48 | |
| 500 g | |
| 635684 | |
| 14,2145 | |
| SSZWWUDQMAHNAQ-UHFFFAOYNA-N | |
| 3-chloropropane-1,2-diol | |
| 7290 | |
| 110.54 |
Chemical Identifiers
| 96-24-2 | |
| 110.54 | |
| SSZWWUDQMAHNAQ-UHFFFAOYNA-N | |
| 7290 | |
| 3-chloropropane-1,2-diol |
| C3H7ClO2 | |
| MFCD00004712 | |
| 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin | |
| CHEBI:18721 | |
| OCC(O)CCl |
Safety and Handling
GHS H Statement
H300-H360-H351-H318-H312-H332-H315-H335
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing cancer.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P271-P280i-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P361-P363-P501c
H300+H330-H311-H315-H319-H335-H340-H350-H360-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: GLYCEROL ALPHA-MONOCHLOROHYDRIN
EINECSNumber : 202-492-4
RTECSNumber : TY4025000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only