missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Chloro-1,2-propanediol, 98%

Catalog No. p-7023080
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1614336 500 g
AAA161430E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1614336 Supplier Thermo Scientific Chemicals Supplier No. A1614336
Only null left

CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-24-2
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.54
MDL Number MFCD00004712
InChI Key SSZWWUDQMAHNAQ-UHFFFAOYNA-N
Synonym 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
PubChem CID 7290
ChEBI CHEBI:18721
IUPAC Name 3-chloropropane-1,2-diol
SMILES OCC(O)CCl

Specifications

Melting Point -40°C
Density 1.322
Boiling Point 213°C
Flash Point 137°C (278°F)
Refractive Index 1.48
Quantity 500 g
UN Number UN2689
Beilstein 635684
Sensitivity Hygroscopic
Merck Index 14,2145
Formula Weight 110.54
Percent Purity 98%
Chemical Name or Material (+/-)-3-Chloro-1,2-propanediol
Show More Show Less
Hazard Category H300+H330-H311-H315-H319-H335-H340-H350-H360-H372
Hazard Statement GHS H Statement
H300-H360-H351-H318-H312-H332-H315-H335
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing cancer.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: GLYCEROL ALPHA-MONOCHLOROHYDRIN
EINECSNumber 202-492-4
RTECSNumber TY4025000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only