missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-3-Chloro-1,2-propane-1,1,2,3,3-d5-diol, MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 342611-01-2 |
| Physical Form | Neat |
| Linear Formula | C3D5H2 ClO2 |
| MDL Number | MFCD01317355 |
| Synonym | 3-MCPD-d5 |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 115.57 |
| Grade | Analytical Standard |