missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2,4-triazole 98.0+%, TCI America™

Catalog No. C30131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30131G 1 g
C30135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30131G Supplier TCI America Supplier No. C30131G

Chemical Identifiers

CAS 6818-99-1
Molecular Formula C2H2ClN3
Molecular Weight (g/mol) 103.51
MDL Number MFCD00128812
InChI Key QGOUKZPSCTVYLX-UHFFFAOYSA-N
Synonym 3-chloro-1,2,4-triazole, 3-chloro-1h-1,2,4-triazole, 3-chloro-4h-1,2,4-triazole, 1h-1,2,4-triazole, 3-chloro, 3-chloro-s-triazole, s-triazole, 3-chloro, 1h-1,2,4-triazole, 5-chloro, 3-chloro-1h-1,2,4 triazole, 3-chloro-2h-1,2,4-triazole
PubChem CID 81265
IUPAC Name 5-chloro-1H-1,2,4-triazole
SMILES ClC1=NC=NN1

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 1 g
Formula Weight 103.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-1,2,4-triazole