accessibility menu, dialog, popup

UserName

ClNHNN

CAS RN 6818-99-1

IUPAC Name: 5-chloro-1H-1,2,4-triazole
Synonyms: 3-chloro-2H-1,2,4-triazole
Molecular Weight (g/mol): 103.51
Molecular Formula: C2H2ClN3
InChi Key: QGOUKZPSCTVYLX-UHFFFAOYSA-N
SMILES: ClC1=NC=NN1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)