missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2,4-triazole, 97+%, Thermo Scientific™

Catalog No. AAH5238703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5238703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5238703 Supplier Thermo Scientific Chemicals Supplier No. H5238703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-1,2,4-triazole is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 6818-99-1
Molecular Formula C2H2ClN3
Molecular Weight (g/mol) 103.51
MDL Number MFCD00128812
InChI Key QGOUKZPSCTVYLX-UHFFFAOYSA-N
Synonym 3-chloro-1,2,4-triazole, 3-chloro-1h-1,2,4-triazole, 3-chloro-4h-1,2,4-triazole, 1h-1,2,4-triazole, 3-chloro, 3-chloro-s-triazole, s-triazole, 3-chloro, 1h-1,2,4-triazole, 5-chloro, 3-chloro-1h-1,2,4 triazole, 3-chloro-2h-1,2,4-triazole
PubChem CID 81265
IUPAC Name 5-chloro-1H-1,2,4-triazole
SMILES ClC1=NC=NN1

Specifications

Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 103.5
Percent Purity ≥97%
Chemical Name or Material 3-Chloro-1,2,4-triazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-892-1
TSCA Yes
Recommended Storage Ambient temperatures