Learn More
(+)-3-Carene, Thermo Scientific™
Chemical Identifiers
| CAS | 498-15-7 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00066417 |
| InChI Key | BQOFWKZOCNGFEC-BDAKNGLRSA-N |
| IUPAC Name | (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene |
| SMILES | CC1=CC[C@@H]2[C@H](C1)C2(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC463990050
|
Thermo Scientific Chemicals
463990050 |
5 mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC463990250
|
Thermo Scientific Chemicals
463990250 |
25 mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 498-15-7 | |
| 136.24 | |
| BQOFWKZOCNGFEC-BDAKNGLRSA-N | |
| CC1=CC[C@@H]2[C@H](C1)C2(C)C |
| C10H16 | |
| MFCD00066417 | |
| (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene |
Specifications
| 498-15-7 | |
| 170.0°C | |
| C10H16 | |
| 5 mL | |
| Solubility aq. soln.: .. Other solubilities: soluble in benzene, acetone, ethanol | |
| CC1=CC[C@@H]2[C@H](C1)C2(C)C | |
| 136.24 | |
| (+)-3-Carene |
| 0.8650 g/mL | |
| 46°C | |
| MFCD00066417 | |
| 0.865 | |
| BQOFWKZOCNGFEC-BDAKNGLRSA-N | |
| (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene | |
| 136.23 |
Safety and Handling
GHS H Statement Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water or shower IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing.
Wash face, hands and any exposed skin thoroughly after handling.
If skin irritation or rash occurs: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 207-856-6
RUO â Research Use Only