missing translation for 'onlineSavingsMsg'
Learn More

3-Carene, 90%, stabilized

CAS: 13466-78-9 | C10H16 | 136.23 g/mol

$52.88 - $188.67

Chemical Identifiers

CAS 13466-78-9 , 138-86-3 , 127-91-3 , 99-86-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.23
MDL Number MFCD00001315
InChI Key BQOFWKZOCNGFEC-UHFFFAOYSA-N
Synonym 3-carene, carene, 3,7,7-trimethylbicyclo 4.1.0 hept-3-ene, delta-3-carene, monoterpenes, alpha-carene, bicyclo 4.1.0 hept-3-ene, 3,7,7-trimethyl, +--delta3-carene, + car-3-ene, +--3-carene
PubChem CID 26049
ChEBI CHEBI:35661
IUPAC Name 4,7,7-trimethylbicyclo[4.1.0]hept-3-ene
SMILES CC1=CCC2C(C1)C2(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC367960250
View Documents Promotion Details
N/A 25 mL Glass bottle
Each for $52.88
Only null left
 
AC367960010
View Documents Promotion Details
N/A 1 L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13466-78-9 , 138-86-3 , 127-91-3 , 99-86-5
136.23
BQOFWKZOCNGFEC-UHFFFAOYSA-N
26049
4,7,7-trimethylbicyclo[4.1.0]hept-3-ene
C10H16
MFCD00001315
3-carene, carene, 3,7,7-trimethylbicyclo 4.1.0 hept-3-ene, delta-3-carene, monoterpenes, alpha-carene, bicyclo 4.1.0 hept-3-ene, 3,7,7-trimethyl, +--delta3-carene, + car-3-ene, +--3-carene
CHEBI:35661
CC1=CCC2C(C1)C2(C)C

Specifications

13466-78-9 , 138-86-3 , 127-91-3 , 99-86-5
100.0
0.8640g/mL
45°C
88% min. (GC)
1.4720 to 1.474
05, 94
15, 1835
Solubility in water: insoluble
CC1=CCC2C(C1)C2(C)C
136.23
26049
136.23
Liquid
88.0
Colorless to Yellow
166°C to 170°C
Authentic
C10H16
MFCD00001315
0.864
3-carene, carene, 3,7,7-trimethylbicyclo 4.1.0 hept-3-ene, delta-3-carene, monoterpenes, alpha-carene, bicyclo 4.1.0 hept-3-ene, 3,7,7-trimethyl, +--delta3-carene, + car-3-ene, +--3-carene
BQOFWKZOCNGFEC-UHFFFAOYSA-N
4,7,7-trimethylbicyclo[4.1.0]hept-3-ene
0.01% alpha-Tocopherol
CHEBI:35661
90%
3-Carene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Very toxic to aquatic life with long l

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 236-719-3

SDS
Documents

Documents

RUO – Research Use Only