missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-5-nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C23851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23851G Supplier TCI America Supplier No. C23851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Carboxy-5-nitrophenylboronic Acid (contains varying amounts of Anhydride)
CAS 101084-81-5
Melting Point 229°C
Color White-Yellow
Molecular Formula C7H6BNO6
Quantity 1 g
MDL Number MFCD00757433
Synonym 3-carboxy-5-nitrophenylboronic acid, 3-nitro-5-carboxyphenylboronic acid, 5-nitro-3-carboxyphenylboronic acid, 3-carboxy-5-nitrophenyl boronic acid, 3-carboxy-5-nitrobenzeneboronic acid, benzoic acid, 3-borono-5-nitro, 3-dihydroxyboryl-5-nitrobenzoic acid, 3-dihydroxyboranyl-5-nitrobenzoic acid, 3-borono-5-nitro-benzoic acid, pubchem5139
InChI Key WNIFCLWDGNHGMX-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O)(O)O
IUPAC Name 3-borono-5-nitrobenzoic acid
Molecular Weight (g/mol) 210.936
PubChem CID 2773315
Formula Weight 210.94
Physical Form Crystalline Powder
Show More Show Less