missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-5-nitrobenzeneboronic acid, 97%

CAS: 101084-81-5 | C7H6BNO6 | 210.936 g/mol

$114.38 - $347.77

Chemical Identifiers

CAS 101084-81-5
Molecular Formula C7H6BNO6
Molecular Weight (g/mol) 210.936
MDL Number MFCD00757433
InChI Key WNIFCLWDGNHGMX-UHFFFAOYSA-N
Synonym 3-carboxy-5-nitrophenylboronic acid, 3-nitro-5-carboxyphenylboronic acid, 5-nitro-3-carboxyphenylboronic acid, 3-carboxy-5-nitrophenyl boronic acid, 3-carboxy-5-nitrobenzeneboronic acid, benzoic acid, 3-borono-5-nitro, 3-dihydroxyboryl-5-nitrobenzoic acid, 3-dihydroxyboranyl-5-nitrobenzoic acid, 3-borono-5-nitro-benzoic acid, pubchem5139
PubChem CID 2773315
IUPAC Name 3-borono-5-nitrobenzoic acid
SMILES B(C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2934803
View Documents
Thermo Scientific Chemicals
H2934803
1 g
Each for $114.38
Only null left
 
AAH2934806
View Documents
Thermo Scientific Chemicals
H2934806
5 g
Each for $347.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

101084-81-5
210.936
WNIFCLWDGNHGMX-UHFFFAOYSA-N
2773315
B(C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O)(O)O
C7H6BNO6
MFCD00757433
3-carboxy-5-nitrophenylboronic acid, 3-nitro-5-carboxyphenylboronic acid, 5-nitro-3-carboxyphenylboronic acid, 3-carboxy-5-nitrophenyl boronic acid, 3-carboxy-5-nitrobenzeneboronic acid, benzoic acid, 3-borono-5-nitro, 3-dihydroxyboryl-5-nitrobenzoic acid, 3-dihydroxyboranyl-5-nitrobenzoic acid, 3-borono-5-nitro-benzoic acid, pubchem5139
3-borono-5-nitrobenzoic acid

Specifications

101084-81-5
MFCD00757433
3-carboxy-5-nitrophenylboronic acid, 3-nitro-5-carboxyphenylboronic acid, 5-nitro-3-carboxyphenylboronic acid, 3-carboxy-5-nitrophenyl boronic acid, 3-carboxy-5-nitrobenzeneboronic acid, benzoic acid, 3-borono-5-nitro, 3-dihydroxyboryl-5-nitrobenzoic acid, 3-dihydroxyboranyl-5-nitrobenzoic acid, 3-borono-5-nitro-benzoic acid, pubchem5139
B(C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O)(O)O
210.936
210.94
3-Carboxy-5-nitrobenzeneboronic acid
C7H6BNO6
1 g
WNIFCLWDGNHGMX-UHFFFAOYSA-N
3-borono-5-nitrobenzoic acid
2773315
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only