missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Carbamoylphenylboronic acid pinacol ester, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 444450050
| Quantity | 5g |
|---|
Chemical Identifiers
| 188665-74-9 | |
| 247.1 | |
| 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-aminocarbonylphenylboronic acid, pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzamide, 3-aminocarbonylphenylboronic acid pinacol ester, benzamide, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 2-dioxaborolan-2-yl-benzamide, bpkiphywhvowms-uhfffaoysa-n, 3-carbamoylphenylboronic acid pinacol ester |
| C13H18BNO3 | |
| BPKIPHYWHVOWMS-UHFFFAOYSA-N | |
| 2760587 |
Specifications
| 3-Carbamoylphenylboronic acid pinacol ester | |
| 96% min. (GC) | |
| 5g | |
| BPKIPHYWHVOWMS-UHFFFAOYSA-N | |
| 2760587 | |
| 97% |
| 188665-74-9 | |
| C13H18BNO3 | |
| 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-aminocarbonylphenylboronic acid, pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzamide, 3-aminocarbonylphenylboronic acid pinacol ester, benzamide, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 2-dioxaborolan-2-yl-benzamide, bpkiphywhvowms-uhfffaoysa-n, 3-carbamoylphenylboronic acid pinacol ester | |
| 247.1 | |
| 247.1 |
Safety and Handling
GHS Signal Word: Warning