accessibility menu, dialog, popup

UserName

CAS RN 188665-74-9

CH3H3COBOH3CH3CNH2O

CAS RN 188665-74-9

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 247.1
Molecular Formula: C13H18BNO3
InChi Key: BPKIPHYWHVOWMS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)