missing translation for 'onlineSavingsMsg'
Learn More

3-Carbamoyl-4-fluorobenzeneboronic acid, 97%

Catalog No. p-7035385
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53102MD 250 mg
AAH5310203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53102MD Supplier Thermo Scientific Chemicals Supplier No. H53102MD

CAS: 874219-34-8 | C7H7BFNO3 | 182.945 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874219-34-8
Molecular Formula C7H7BFNO3
Molecular Weight (g/mol) 182.945
MDL Number MFCD08235053
InChI Key RNNYXSWJFLHIRC-UHFFFAOYSA-N
Synonym 3-carbamoyl-4-fluorophenyl boronic acid, 3-carbamoyl-4-fluorobenzeneboronic acid, 3-aminocarbonyl-4-fluorophenylboronic acid, 3-aminocarbonyl-4-fluorobenzeneboronic acid, acmc-209qky, 3-carbamoyl-4-fluorophenyl boronicacid, 4-fluoro-3-carbamoyl phenylboronic acid, 3-aminocarbonyl-4-fluoranyl-phenyl boronic acid, boronic acid,b-3-aminocarbonyl-4-fluorophenyl
PubChem CID 44717530
IUPAC Name (3-carbamoyl-4-fluorophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)C(=O)N)(O)O

Specifications

Melting Point 257°C to 259°C
Quantity 250 mg
Formula Weight 182.95
Percent Purity 97%
Chemical Name or Material 3-Carbamoyl-4-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only