missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-2-one 97.0+%, TCI America™

Catalog No. B23931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23931G 1 g
B23935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23931G Supplier TCI America Supplier No. B23931G

Chemical Identifiers

CAS 1423-60-5
Molecular Formula C4H4O
Molecular Weight (g/mol) 68.08
MDL Number MFCD00008775
InChI Key XRGPFNGLRSIPSA-UHFFFAOYSA-N
Synonym 3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
PubChem CID 15018
ChEBI CHEBI:48060
IUPAC Name but-3-yn-2-one
SMILES CC(=O)C#C

Specifications

Color Yellow
Boiling Point 86°C
Quantity 1 g
UN Number 1992
Formula Weight 68.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Butyn-2-one
RTECSNumber ES0875000
TSCA No
Recommended Storage Refrigerator