accessibility menu, dialog, popup

UserName

CAS RN 1423-60-5

IUPAC Name: but-3-yn-2-one
Synonyms: butyn-2-one
Molecular Weight (g/mol): 68.08
Molecular Formula: C4H4O
InChi Key: XRGPFNGLRSIPSA-UHFFFAOYSA-N
SMILES: CC(=O)C#C
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)