missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-2-one, 98%

CAS: 1423-60-5 | C4H4O | 68.08 g/mol

Supplier:  Thermo Scientific Chemicals L0552706

 View more versions of this product

Catalog No. AAL0552706

Only null left
Explore available promotions

Description

Description

3-Butyn-2-one was used in the synthesis of clerodane diterpenoid (±)-sacacarin. It was used as substrate in stereoselective, conjugate arylation mediated by gallium(III) chloride leading to (E)-α,β-unsaturated ketones. 3-Butyn-2-one undergoes asymmetric double-Michael reaction with ortho-tosylamidophenyl malonate catalyzed by chiral aminophosphines to yield indolines2. It undergoes double Michael reaction with nitrogen-containing tethered diacid to give pipecolic acid derivatives. 3-Butyn-2-one, is used as a reactant with Dimethyl­ acetone-1,3-dicarboxyl­ate, under mild conditions to give a benzenoid product via a Michael addition - aldol cyclization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Butyn-2-one was used in the synthesis of clerodane diterpenoid (+/-)-sacacarin. It was used as substrate in stereoselective, conjugate arylation mediated by gallium(III) chloride leading to (E)-α,β-unsaturated ketones. 3-Butyn-2-one undergoes asymmetric double-Michael reaction with ortho-tosylamidophenyl malonate catalyzed by chiral aminophosphines to yield indolines2. It undergoes double Michael reaction with nitrogen-containing tethered diacid to give pipecolic acid derivatives. 3-Butyn-2-one, is used as a reactant with Dimethyl­ acetone-1,3-dicarboxyl­ate, under mild conditions to give a benzenoid product via a Michael addition - aldol cyclization process.

Solubility
It is soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications

Specifications

3-Butyn-2-one
0.87
−1°C (30°F)
1.406
5 g
605353
It is soluble in water.
CC(=O)C#C
68.08
CHEBI:48060
98%
1423-60-5
84°C to 86°C
C4H4O
MFCD00008775
UN1992
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
XRGPFNGLRSIPSA-UHFFFAOYSA-N
but-3-yn-2-one
15018
68.08

Chemical Identifiers

1423-60-5
68.08
XRGPFNGLRSIPSA-UHFFFAOYSA-N
15018
but-3-yn-2-one
C4H4O
MFCD00008775
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
CHEBI:48060
CC(=O)C#C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H318-H315-H335
Highly flammable liquid and vapor.
Fatal if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H300-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 215-834-2

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only