missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-2-one, 98%

Catalog No. AAL0552703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0552703 1 g
AAL0552706 5 g
AAL0552714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0552703 Supplier Thermo Scientific Chemicals Supplier No. L0552703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1423-60-5 | C4H4O | 68.08 g/mol

3-Butyn-2-one was used in the synthesis of clerodane diterpenoid (±)-sacacarin. It was used as substrate in stereoselective, conjugate arylation mediated by gallium(III) chloride leading to (E)-α,β-unsaturated ketones. 3-Butyn-2-one undergoes asymmetric double-Michael reaction with ortho-tosylamidophenyl malonate catalyzed by chiral aminophosphines to yield indolines2. It undergoes double Michael reaction with nitrogen-containing tethered diacid to give pipecolic acid derivatives. 3-Butyn-2-one, is used as a reactant with Dimethyl­ acetone-1,3-dicarboxyl­ate, under mild conditions to give a benzenoid product via a Michael addition - aldol cyclization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Butyn-2-one was used in the synthesis of clerodane diterpenoid (+/-)-sacacarin. It was used as substrate in stereoselective, conjugate arylation mediated by gallium(III) chloride leading to (E)-α,β-unsaturated ketones. 3-Butyn-2-one undergoes asymmetric double-Michael reaction with ortho-tosylamidophenyl malonate catalyzed by chiral aminophosphines to yield indolines2. It undergoes double Michael reaction with nitrogen-containing tethered diacid to give pipecolic acid derivatives. 3-Butyn-2-one, is used as a reactant with Dimethyl­ acetone-1,3-dicarboxyl­ate, under mild conditions to give a benzenoid product via a Michael addition - aldol cyclization process.

Solubility
It is soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 1423-60-5
Molecular Formula C4H4O
Molecular Weight (g/mol) 68.08
MDL Number MFCD00008775
InChI Key XRGPFNGLRSIPSA-UHFFFAOYSA-N
Synonym 3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
PubChem CID 15018
ChEBI CHEBI:48060
IUPAC Name but-3-yn-2-one
SMILES CC(=O)C#C

Specifications

Density 0.87
Boiling Point 84°C to 86°C
Flash Point −1°C (30°F)
Refractive Index 1.406
Quantity 1 g
UN Number UN1992
Beilstein 605353
Solubility Information It is soluble in water.
Formula Weight 68.08
Percent Purity 98%
Chemical Name or Material 3-Butyn-2-one
Show More Show Less
Hazard Category H225-H300-H315-H319-H335
Hazard Statement GHS H Statement
H225-H300-H318-H315-H335
Highly flammable liquid and vapor.
Fatal if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 215-834-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only