missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-2-one, 96%, Thermo Scientific™

Chemical Identifiers

CAS 1423-60-5
Molecular Formula C4H4O
Molecular Weight (g/mol) 68.08
MDL Number MFCD00008775
InChI Key XRGPFNGLRSIPSA-UHFFFAOYSA-N
Synonym 3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
PubChem CID 15018
ChEBI CHEBI:48060
IUPAC Name but-3-yn-2-one
SMILES CC(=O)C#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC330440025
SDS View Documents
Thermo Scientific Chemicals
330440025
2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC330440100
SDS View Documents
Thermo Scientific Chemicals
330440100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1423-60-5
68.08
XRGPFNGLRSIPSA-UHFFFAOYSA-N
15018
but-3-yn-2-one
C4H4O
MFCD00008775
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
CHEBI:48060
CC(=O)C#C

Specifications

1423-60-5
85.0°C
Authentic
Glass bottle
1.4045 to 1.4095
MFCD00008775
03,38
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
CC(=O)C#C
68.08
CHEBI:48060
96%
0.8700g/mL
−1°C
95% min. (GC)
C4H4O
HC≡CCOCH3
2.5g
0.87
XRGPFNGLRSIPSA-UHFFFAOYSA-N
but-3-yn-2-one
15018
68.06
3-Butyn-2-one
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 215-834-2

SDS
Documents

Documents

RUO – Research Use Only