missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-2-ol 97.0+%, TCI America™

Catalog No. p-7118795
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
B10015ML 5 mL
B100125ML 25 mL
2 options

Catalog No. B10015ML

Supplier: TCI America B10015ML

Only null left

Chemical Identifiers

CAS 2028-63-9
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00004541
InChI Key GKPOMITUDGXOSB-UHFFFAOYNA-N
Synonym 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol
PubChem CID 16239
IUPAC Name but-3-yn-2-ol
SMILES CC(O)C#C

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 mL
UN Number 2929
Formula Weight 70.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Butyn-2-ol
RTECSNumber ES0709800
TSCA Yes