accessibility menu, dialog, popup

UserName

CAS RN 2028-63-9

IUPAC Name: but-3-yn-2-ol
Synonyms: 3-butyn-2-ol
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: GKPOMITUDGXOSB-UHFFFAOYNA-N
SMILES: CC(O)C#C
Boiling Point 
  • (5)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)