Learn More
(±)-3-Butyn-2-ol, 98%
CAS: 2028-63-9 | C4H6O | 70.09 g/mol
Supplier: Thermo Scientific Chemicals A1188122
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (±)-3-Butyn-2-ol | |
| 0.894 | |
| 28°C (82°F) | |
| 1.426 | |
| 100 g | |
| 635722 | |
| GKPOMITUDGXOSB-UHFFFAOYNA-N | |
| but-3-yn-2-ol | |
| 16239 | |
| 98% |
| 2028-63-9 | |
| 108°C to 109°C | |
| C4H6O | |
| MFCD00004541 | |
| UN2929 | |
| 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol | |
| CC(O)C#C | |
| 70.09 | |
| 70.09 |
Chemical Identifiers
| 2028-63-9 | |
| 70.09 | |
| GKPOMITUDGXOSB-UHFFFAOYNA-N | |
| 16239 | |
| CC(O)C#C |
| C4H6O | |
| MFCD00004541 | |
| 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol | |
| but-3-yn-2-ol |
Safety and Handling
GHS H Statement
H300-H310-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H300+H310+H330-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber : 217-978-1
RTECSNumber : ES0709800
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only