Learn More
3-Butyn-2-ol, 97%, Thermo Scientific™
$78.05 - $299.24
Chemical Identifiers
| CAS | 2028-63-9 |
|---|---|
| Molecular Formula | C4H6O |
| Molecular Weight (g/mol) | 70.09 |
| MDL Number | MFCD00004541 |
| InChI Key | GKPOMITUDGXOSB-UHFFFAOYNA-N |
| Synonym | 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol |
| PubChem CID | 16239 |
| IUPAC Name | but-3-yn-2-ol |
| SMILES | CC(O)C#C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC245310100
|
N/A | 10g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC245310500
|
N/A | 50g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C4H6O | |
| MFCD00004541 | |
| 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol | |
| but-3-yn-2-ol |
Specifications
| 2028-63-9 | |
| 100.0 | |
| 108°C to 111°C | |
| Authentic | |
| C4H6O | |
| MFCD00004541 | |
| 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol | |
| GKPOMITUDGXOSB-UHFFFAOYNA-N | |
| but-3-yn-2-ol | |
| 16239 | |
| 97% |
| 96.0 | |
| 0.8900g/mL | |
| 26°C | |
| 96% min. (GC) | |
| 1.4250 to 1.427 | |
| 0.89 | |
| Solubility in water: miscible. Other solubilities: miscible with methanol | |
| CC(O)C#C | |
| 70.09 | |
| 70.09 | |
| 3-Butyn-2-ol |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
May cause respiratory irritation.
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Fatal if inhaled.
GHS P Statement
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash wi
GHS Signal Word: Danger
EINECSNumber : 217-978-1
RUO – Research Use Only