missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-1-ol 97.0+%, TCI America™

Supplier:  TCI America B079925ML

 View more versions of this product

Catalog No. B079925ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Butyn-1-ol
-64°C
130°C
MFCD00002955
1987
OTJZCIYGRUNXTP-UHFFFAOYSA-N
but-3-yn-1-ol
13566
70.09
Liquid
927-74-2
Yellow
C4H6O
25 mL
3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
C#CCCO
70.091
CHEBI:27444
≥97.0% (GC)

Chemical Identifiers

927-74-2
70.091
OTJZCIYGRUNXTP-UHFFFAOYSA-N
13566
but-3-yn-1-ol
C4H6O
MFCD00002955
3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
CHEBI:27444
C#CCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : ES0710000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions