missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-1-ol, 98%

CAS: 927-74-2 | C4H6O | 70.091 g/mol

Supplier:  Thermo Scientific Chemicals A1147714

 View more versions of this product

Catalog No. AAA1147714

Only null left
Explore available promotions

Description

Description

3-butyn-1-ol is used in the study of a palladium-catalyzed coupling with beta-tetrionic acid bromide to prepare alkynyl substituted furanones. It is also used to prepare Markovnikov addition product. Further, it is used as an intermediate of an active pharmaceutical ingredient such as fexofenadine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-butyn-1-ol is used in the study of a palladium-catalyzed coupling with beta-tetrionic acid bromide to prepare alkynyl substituted furanones. It is also used to prepare Markovnikov addition product. Further, it is used as an intermediate of an active pharmaceutical ingredient such as fexofenadine.

Solubility
Miscible with alcohols and organic solvents. Immiscible with aliphatic hydrocarbon solvents.

Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides and copper.
Specifications

Specifications

3-Butyn-1-ol
-63°C
129°C to 130°C
Characteristic
1.4405
MFCD00002955
UN1987
3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
OTJZCIYGRUNXTP-UHFFFAOYSA-N
but-3-yn-1-ol
13566
70.09
927-74-2
0.927
36°C (96°F)
C4H6O
HC≡CCH2CH2OH
25 g
773710
Miscible with alcohols and organic solvents. Immiscible with aliphatic hydrocarbon solvents.
C#CCCO
70.091
CHEBI:27444
≥98%

Chemical Identifiers

927-74-2
70.091
OTJZCIYGRUNXTP-UHFFFAOYSA-N
13566
but-3-yn-1-ol
C4H6O
MFCD00002955
3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
CHEBI:27444
C#CCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 213-161-9

RTECSNumber : ES0710000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only