missing translation for 'onlineSavingsMsg'
Learn More

3-Butylpyridine 99.0+%, TCI America™

Catalog No. b11935g
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
B11935G 5 g
1 options

Catalog No. B11935G

Supplier: TCI America B11935G

Only null left

Chemical Identifiers

CAS 539-32-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00010709
InChI Key QSNMFWFDOFQASV-UHFFFAOYSA-N
PubChem CID 10874
IUPAC Name 3-butylpyridine
SMILES CCCCC1=CC=CN=C1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
Formula Weight 135.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Butylpyridine
RTECSNumber US4056000
TSCA Yes