accessibility menu, dialog, popup

UserName

H3CN

CAS RN 539-32-2

IUPAC Name: 3-butylpyridine
Synonyms: pyridine, 3-butyl-
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: QSNMFWFDOFQASV-UHFFFAOYSA-N
SMILES: CCCCC1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results