Learn More
3-Buten-2-ol, 97%
Important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field. | CAS: 598-32-3 | C4H8O | 72.11 g/mol
Supplier: Thermo Scientific Chemicals L1478706
Description
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Buten-2-ol | |
| <-100°C | |
| 96°C to 98°C | |
| Glass bottle | |
| 1.414 | |
| 5 g | |
| 1361410 | |
| 3-buten-2-ol, 1-buten-3-ol, methyl vinylcarbinol, 3-butene-2-ol, 3-hydroxy-1-butene, propenol, 1-methyl, methyl vinyl carbinol, 1-methyl-2-propenol, 2-methyl-2-propenol, 1-methylallyl alcohol | |
| MKUWVMRNQOOSAT-UHFFFAOYNA-N | |
| but-3-en-2-ol | |
| 11716 | |
| 97% |
| 598-32-3 | |
| 0.838 | |
| 20°C (68°F) | |
| C4H8O | |
| MFCD00004543 | |
| UN1986 | |
| 0.838 | |
| Fully miscible with water. | |
| CC(O)C=C | |
| 72.11 | |
| 72.11 |
Chemical Identifiers
| 598-32-3 | |
| 72.11 | |
| MKUWVMRNQOOSAT-UHFFFAOYNA-N | |
| 11716 | |
| CC(O)C=C |
| C4H8O | |
| MFCD00004543 | |
| 3-buten-2-ol, 1-buten-3-ol, methyl vinylcarbinol, 3-butene-2-ol, 3-hydroxy-1-butene, propenol, 1-methyl, methyl vinyl carbinol, 1-methyl-2-propenol, 2-methyl-2-propenol, 1-methylallyl alcohol | |
| but-3-en-2-ol |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H332-H335
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 209-929-8
RTECSNumber : EM9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only